N-(2-Chloro-6-methylphenyl)-2-((2-methyl-6-(piperazin-1-yl)pyrimidin-4-yl)amino)thiazole-5-carboxamide 98%

Supplier: Ambeed
Danger

A245274-50MG A245274-100MG A245274-1G A245274-250MG
77568-384EA 182 USD
77568-384 77748-439 77748-440 77748-441
N-(2-Chloro-6-methylphenyl)-2-((2-methyl-6-(piperazin-1-yl)pyrimidin-4-yl)amino)thiazole-5-carboxamide 98%
N-(2-Chloro-6-methylphenyl)-2-((2-methyl-6-(piperazin-1-yl)pyrimidin-4-yl)amino)thiazole-5-carboxamide

Precaución: For research use only.

Formula: C₂₀H₂₂ClN₇OS
MDL Number: MFCD18252607
CAS Number: 910297-51-7
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR