(1S)-5,5',6,6',7,7',8,8'-Octahydro-[1,1'-Binaphthalene]-2,2'-diamine 97%

Supplier: Ambeed
Warning

A243923-1G A243923-250MG
77568-370EA 94.76 USD
77568-370 77600-968
(1S)-5,5',6,6',7,7',8,8'-Octahydro-[1,1'-Binaphthalene]-2,2'-diamine 97%
(1S)-5,5',6,6',7,7',8,8'-Octahydro-[1,1'-Binaphthalene]-2,2'-diamine

Precaución: For research use only.

Formula: C₂₀H₂₄N₂
MW: 292.42 g/mol
MDL Number: MFCD19443506
CAS Number: 229177-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR