2,2-Bis(4-carboxyphenyl)hexafluoropropane 98%

Supplier: Ambeed
Warning

Synonyms: 4,4'-(Hexafluoroisopropylidene)bis(benzoic Acid)

A242950-5G A242950-1G A242950-25G
77568-358EA 44 USD
77568-358 77881-640 77881-641
2,2-Bis(4-carboxyphenyl)hexafluoropropane 98%
2,2-Bis(4-carboxyphenyl)hexafluoropropane

Precaución: For research use only.

Formula: C₁₇H₁₀F₆O₄
MW: 392.25 g/mol
MDL Number: MFCD00040931
CAS Number: 1171-47-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR