Benzo[d]thiazol-2-ylmethanamine 95%, Ambeed.Inc

Supplier: Ambeed
Danger

A238756-1G A238756-250MG A238756-100MG
77568-320EA 174 USD
77568-320 77683-721 77885-616
Benzo[d]thiazol-2-ylmethanamine 95%, Ambeed.Inc
Benzo[d]thiazol-2-ylmethanamine

Precaución: For research use only.

Formula: C₈H₈N₂S
MDL Number: MFCD06660504
CAS Number: 42182-65-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR