2,6-Dichloro-4-fluoropyridine 95%

Supplier: Ambeed
Danger

A238692-1G A238692-5G
77568-312EA 210.12 USD
77568-312 77568-314
2,6-Dichloro-4-fluoropyridine 95%
2,6-Dichloro-4-fluoropyridine

Precaución: For research use only.

Formula: C₅H₂Cl₂FN
MW: 165.981 g/mol
MDL Number: MFCD13185534
CAS Number: 52074-49-4
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR