1H-Indole-3-boronic acid, pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole, 2-(1H-Indol-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A238667-1G A238667-250MG A238667-5G
77568-310EA 126 USD
77568-310 77612-316 77883-633
1H-Indole-3-boronic acid, pinacol ester 98%
1H-Indole-3-boronic acid, pinacol ester

Precaución: For research use only.

Formula: C₁₄H₁₈BNO₂
MW: 243.109 g/mol
MDL Number: MFCD09879158
CAS Number: 937366-54-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR