Chlorobis(ethylene)rhodium(I) dimer 95%

Supplier: Ambeed
Warning

Synonyms: Bis(ethylene)rhodium(I) chloride dimer, [Rh(C₂H₄)₂Cl]₂, µ-Dichlorotetraethylene dirhodium(I), Dichlorotetra(ethylene)dirhodium(I), Dichlorotetrakis(ethylene)dirhodium(I)

A233945-100MG A233945-250MG A233945-1G A233945-50MG
77568-268EA 81 USD
77568-268 77568-270 77600-876 77600-878
Chlorobis(ethylene)rhodium(I) dimer 95%
Chlorobis(ethylene)rhodium(I) dimer

Precaución: For research use only.

Formula: C₈H₁₆Cl₂Rh₂
MDL Number: MFCD00013206
CAS Number: 12081-16-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR