N-tert-Butyltrimethylsilylamine 95%

Supplier: Ambeed
Danger

A232107-25G A232107-5G
77568-248EA 52 USD
77568-248 77568-250
N-tert-Butyltrimethylsilylamine 95%
N-tert-Butyltrimethylsilylamine

Precaución: For research use only.

Formula: C₇H₁₉NSi
MDL Number: MFCD00054891
CAS Number: 5577-67-3
UN: 2733
ADR: 3,8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR