(S)-3-Amino-1-azabicyclo[2.2.2]octane 98%

Supplier: Ambeed
Danger

A231188-1G A231188-100MG A231188-250MG
77568-238EA 397.58 USD
77568-238 77612-252 77612-254
(S)-3-Amino-1-azabicyclo[2.2.2]octane 98%
(S)-3-Amino-1-azabicyclo[2.2.2]octane

Precaución: For research use only.

Formula: C₇H₁₄N₂
MW: 126.19 g/mol
MDL Number: MFCD08669631
CAS Number: 120570-05-0
UN: 3259
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR