Nickamine 98%

Supplier: Ambeed
Danger

Synonyms: Bis[(2-dimethylamino)phenyl]amine nickel(II) chloride

A226756-250MG A226756-100MG
77568-194EA 282 USD
77568-194 77685-620
Nickamine 98%
Nickamine

Precaución: For research use only.

Formula: C₁₆H₂₀ClN₃Ni
MW: 348.5 g/mol
MDL Number: MFCD17015253
CAS Number: 1033772-47-2
UN: 3077
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR