6-(2,4-Dichlorophenyl)-8-ethyl-2-((3-fluoro-4-(piperazin-1-yl)phenyl)amino)pyrido[2,3-d]pyrimidin-7(8H)-one 98+%

Supplier: Ambeed
Warning

Synonyms: FRAX486

A225881-50MG A225881-10MG A225881-1MG A225881-5MG
77568-186EA 148.32 USD
77568-186 77615-878 77615-880 77615-882
6-(2,4-Dichlorophenyl)-8-ethyl-2-((3-fluoro-4-(piperazin-1-yl)phenyl)amino)pyrido[2,3-d]pyrimidin-7(8H)-one 98+%
6-(2,4-Dichlorophenyl)-8-ethyl-2-((3-fluoro-4-(piperazin-1-yl)phenyl)amino)pyrido[2,3-d]pyrimidin-7(8H)-one

Precaución: For research use only.

Formula: C₂₅H₂₃Cl₂FN₆O
MW: 513.39 g/mol
MDL Number: MFCD26793866
CAS Number: 1232030-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR