Potassium azetidin-3-yl(trifluoro)boranuide, NBOC protected 98%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: Potassium trifluoro-[1-[(2-methylpropan-2-yl)oxycarbonyl]azetidin-3-yl]boranuide, Potassium (1-(tert-butoxycarbonyl)azetidin-3-yl)trifluoroborate

A223822-1G A223822-100MG A223822-250MG A223822-5G
77568-166EA 198 USD
77568-166 77612-174 77683-651 77683-652
Potassium azetidin-3-yl(trifluoro)boranuide, NBOC protected 98%, Ambeed.Inc
Potassium azetidin-3-yl(trifluoro)boranuide, NBOC protected

Precaución: For research use only.

Formula: C₈H₁₄BF₃NO₂·K
MDL Number: MFCD18712148
CAS Number: 1430219-73-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR