1-Bromo-2-methoxy-4-methyl-5-nitrobenzene 98%

Supplier: Ambeed
Warning

A218458-5G A218458-100MG A218458-1G A218458-250MG
77568-118EA 254 USD
77568-118 77883-555 77883-556 77883-557
1-Bromo-2-methoxy-4-methyl-5-nitrobenzene 98%
1-Bromo-2-methoxy-4-methyl-5-nitrobenzene

Precaución: For research use only.

Formula: C₈H₈BrNO₃
MDL Number: MFCD11007872
CAS Number: 861076-28-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR