α-Bromo-4-(trifluoromethoxy)toluene 97%

Supplier: Ambeed
Danger

Synonyms: 1-(Bromomethyl)-4-(trifluoromethoxy)benzene, p-(Trifluoromethoxy)benzyl bromide, 4-(Trifluoromethoxy)benzyl bromide

A216393-5G A216393-25G
77568-092EA 20.6 USD
77568-092 77685-583
α-Bromo-4-(trifluoromethoxy)toluene 97%
α-Bromo-4-(trifluoromethoxy)toluene

Precaución: For research use only.

Formula: C₈H₆BrF₃O
MW: 255.03 g/mol
MDL Number: MFCD00061238
CAS Number: 50824-05-0
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR