2-Methyl-3-pyridineboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (2-Methylpyridin-3-yl)boronic acid

A212903-1G A212903-250MG
77568-054EA 49 USD
77568-054 77612-094
2-Methyl-3-pyridineboronic acid 98%
2-Methyl-3-pyridineboronic acid

Precaución: For research use only.

Formula: C₆H₈BNO₂
MW: 136.9400 g/mol
MDL Number: MFCD07368825
CAS Number: 899436-71-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR