3-(tert-Butoxycarbonyl)-3-azabicyclo[3.1.0]hexane-1-carboxylic acid 95%

Supplier: Ambeed
Warning

A210802-1G A210802-5G A210802-250MG A210802-100MG
77568-038EA 66 USD
77568-038 77593-374 77685-551 77892-320
3-(tert-Butoxycarbonyl)-3-azabicyclo[3.1.0]hexane-1-carboxylic acid 95%
3-(tert-Butoxycarbonyl)-3-azabicyclo[3.1.0]hexane-1-carboxylic acid

Precaución: For research use only.

Formula: C₁₁H₁₇NO₄
MDL Number: MFCD22566159
CAS Number: 1363381-55-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR