Di(adamantan-1-yl)(benzyl)phosphine 98%

Supplier: Ambeed
Warning

Synonyms: Di(1-adamantyl)benzylphosphine

A207350-1G A207350-5G A207350-250MG A207350-25G
77567-988EA 28 USD
77567-988 77600-676 77626-050 77877-643
Di(adamantan-1-yl)(benzyl)phosphine 98%
Di(adamantan-1-yl)(benzyl)phosphine

Precaución: For research use only.

Formula: C₂₇H₃₇P
MDL Number: MFCD05861603
CAS Number: 395116-70-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR