1-Bromo-3,5-diethylbenzene 98%

Supplier: Ambeed
Warning

A205299-10G A205299-1G A205299-5G
77567-972EA 76 USD
77567-972 77600-672 77879-279
1-Bromo-3,5-diethylbenzene 98%
1-Bromo-3,5-diethylbenzene

Precaución: For research use only.

Formula: C₁₀H₁₃Br
MDL Number: MFCD08062420
CAS Number: 90267-03-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR