(S)-tert-Butyl 2-(((benzyloxy)carbonyl)amino)-3-hydroxypropanoate 95%

Supplier: Ambeed
Warning

A204191-5G A204191-25G
77567-956EA 143 USD
77567-956 77885-479
(S)-tert-Butyl 2-(((benzyloxy)carbonyl)amino)-3-hydroxypropanoate 95%
(S)-tert-Butyl 2-(((benzyloxy)carbonyl)amino)-3-hydroxypropanoate

Precaución: For research use only.

Formula: C₁₅H₂₁NO₅
MDL Number: MFCD03428467
CAS Number: 59859-77-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR