Methyl 2-bromo-4-iodobenzoate 97%

Supplier: Ambeed
Warning

A203839-25G A203839-5G
77567-952EA 169 USD
77567-952 77615-708
Methyl 2-bromo-4-iodobenzoate 97%
Methyl 2-bromo-4-iodobenzoate

Precaución: For research use only.

Formula: C₈H₆BrIO₂
MDL Number: MFCD11870175
CAS Number: 1261588-35-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR