D-(+)-Trehalose 98%

Supplier: Ambeed
Warning

Synonyms: (+)-Trehalose, α,α-Trehalose, (2R,2'R,3S,3'S,4S,4'S,5R,5'R,6R,6'R)-6,6'-Oxybis(2-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol), Trehalose, α-D-Glucopyranosyl-α-D-glucopyranoside, D-(+)-Trehalose Anhydrous, D-Trehalose

A201057-100G A201057-25G A201057-1MG A201057-500G
77567-910EA 114 USD
77567-910 77567-912 77667-668 77872-003
D-(+)-Trehalose 98%
D-(+)-Trehalose

Precaución: For research use only.

Formula: C₁₂H₂₂O₁₁
MDL Number: MFCD00006628
CAS Number: 99-20-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR