4-Bromo-N-(2,6-dioxopiperidin-3-yl)-2-fluorobenzamide 95%

Supplier: Ambeed
Warning

A2004383-250MG A2004383-100MG
77567-888EA 157 USD
77567-888 77598-292
4-Bromo-N-(2,6-dioxopiperidin-3-yl)-2-fluorobenzamide 95%
4-Bromo-N-(2,6-dioxopiperidin-3-yl)-2-fluorobenzamide

Precaución: For research use only.

Formula: C₁₂H₁₀BrFN₂O₃
MDL Number: MFCD23167235
CAS Number: 1491427-78-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR