2,5-Difluorobenzonitrile 98%

Supplier: Ambeed
Warning

A199865-25G A199865-10G A199865-5G A199865-100G
77567-878EA 31 USD
77567-878 77625-986 77625-988 77877-585
2,5-Difluorobenzonitrile 98%
2,5-Difluorobenzonitrile

Precaución: For research use only.

Formula: C₇H₃F₂N
MDL Number: MFCD00001777
CAS Number: 64248-64-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR