cis-5-Norbornene endo-2,3-dicarboxylic anhydride 98%

Supplier: Ambeed
Danger

Synonyms: (3aR,4S,7R,7aS)-rel-3a,4,7,7a-Tetrahydro-4,7-methanoisobenzofuran-1,3-dione, Carbic anhydride

A198434-500G A198434-10G A198434-25G A198434-100G
77567-864EA 32 USD
77567-864 77611-908 77611-910 77671-756
cis-5-Norbornene endo-2,3-dicarboxylic anhydride 98%
cis-5-Norbornene endo-2,3-dicarboxylic anhydride

Precaución: For research use only.

Formula: C₉H₈O₃
MDL Number: MFCD00151106
CAS Number: 129-64-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR