Ethyl 5-hydroxy-6-iodonicotinate 98%

Supplier: Ambeed
Warning

A191659-1G A191659-250MG
77567-784EA 115 USD
77567-784 77885-427
Ethyl 5-hydroxy-6-iodonicotinate 98%
Ethyl 5-hydroxy-6-iodonicotinate

Precaución: For research use only.

Formula: C₈H₈INO₃
MDL Number: MFCD13688989
CAS Number: 59288-40-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR