1-N-Boc-2,3-dihydro-pyrrole 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
N-Boc-2-pyrroline
A189926-1G
A189926-5G
77567-772EA
16.48
USD
77567-772
77567-774
1-N-Boc-2,3-dihydro-pyrrole 98%
1-N-Boc-2,3-dihydro-pyrrole
Precaución: For research use only.
Formula:
C₉H₁₅NO₂ MW: 169.221 g/mol |
MDL Number:
MFCD03839942 CAS Number: 73286-71-2 UN: 2810 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...