1H-Pyrrolo[2,3-b]pyridine-6-carbaldehyde 95%

Supplier: Ambeed
Warning

A189358-5G A189358-250MG A189358-1G A189358-100MG
77567-766EA 662 USD
77567-766 77600-538 77671-682 77881-411
1H-Pyrrolo[2,3-b]pyridine-6-carbaldehyde 95%
1H-Pyrrolo[2,3-b]pyridine-6-carbaldehyde

Precaución: For research use only.

Formula: C₈H₆N₂O
MDL Number: MFCD08689966
CAS Number: 898746-46-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR