Benzo[c][1,2]oxaborole-1,6(3H)-diol 95%

Supplier: Ambeed
Warning

A187301-1G A187301-250MG A187301-100MG
77567-742EA 394 USD
77567-742 77671-654 77883-396
Benzo[c][1,2]oxaborole-1,6(3H)-diol 95%
Benzo[c][1,2]oxaborole-1,6(3H)-diol

Precaución: For research use only.

Formula: C₇H₇BO₃
MDL Number: MFCD22200367
CAS Number: 1196473-37-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR