1-Fluoro-2,4,6-trimethylpyridiniumtrifluoromethanesulfonate 85%

Supplier: Ambeed
Danger

Synonyms: 1-Fluoro-2,4,6-trimethylpyridin-1-ium trifluoromethanesulphonate

A186993-1G A186993-250MG A186993-5G A186993-25G A186993-100MG
77567-730EA 158.62 USD
77567-730 77567-732 77567-734 77615-582 77746-202
1-Fluoro-2,4,6-trimethylpyridiniumtrifluoromethanesulfonate 85%
1-Fluoro-2,4,6-trimethylpyridinium trifluoromethanesulphonate

Precaución: For research use only.

Formula: C₉H₁₁F₄NO₃S
MDL Number: MFCD00067525
CAS Number: 107264-00-6
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR