4-Bromo-5-(trifluoromethyl)-1H-indazole 95%

Supplier: Ambeed
Warning

A179394-1G A179394-5G
77567-646EA 81.37 USD
77567-646 77593-138
4-Bromo-5-(trifluoromethyl)-1H-indazole 95%
4-Bromo-5-(trifluoromethyl)-1H-indazole

Precaución: For research use only.

Formula: C₈H₄BrF₃N₂
MW: 265.03 g/mol
MDL Number: MFCD23713147
CAS Number: 1428234-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR