1H-Pyrazole-5-carboxylic acid 97%

Supplier: Ambeed
Warning

A178853-5G A178853-25G
77567-636EA 30.9 USD
77567-636 77615-504
1H-Pyrazole-5-carboxylic acid 97%
1H-Pyrazole-5-carboxylic acid

Precaución: For research use only.

Formula: C₄H₄N₂O₂
MW: 112.0868 g/mol
MDL Number: MFCD00464252
CAS Number: 797027-83-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR