tert-Butyl 4-((2-(4-fluoro-3-(trifluoromethyl)phenyl)-2-oxoethyl)carbamoyl)piperidine-1-carboxylate 98%

Supplier: Ambeed
Warning

A177892-1G A177892-100MG A177892-250MG
77567-614EA 73 USD
77567-614 77871-890 77871-891
tert-Butyl 4-((2-(4-fluoro-3-(trifluoromethyl)phenyl)-2-oxoethyl)carbamoyl)piperidine-1-carboxylate 98%
tert-Butyl 4-((2-(4-fluoro-3-(trifluoromethyl)phenyl)-2-oxoethyl)carbamoyl)piperidine-1-carboxylate

Precaución: For research use only.

Formula: C₂₀H₂₄F₄N₂O₄
MDL Number: MFCD15071415
CAS Number: 1082949-99-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR