4-(1H-Imidazol-1-yl)benzoic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-(1-Imidazolyl)benzoic acid

A175953-5G A175953-250MG
77567-600EA 72.1 USD
77567-600 77600-426
4-(1H-Imidazol-1-yl)benzoic acid 98%
4-(1H-Imidazol-1-yl)benzoic acid

Precaución: For research use only.

Formula: C₁₀H₈N₂O₂
MW: 188.19 g/mol
MDL Number: MFCD00586645
CAS Number: 17616-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR