Imidazo[1,2-a]pyridine-6-boronic acid 97%

Supplier: Ambeed
Warning

A175402-1G A175402-250MG A175402-100MG A175402-5G
77567-590EA 328.57 USD
77567-590 77567-592 77593-106 77683-410
Imidazo[1,2-a]pyridine-6-boronic acid 97%
Imidazo[1,2-a]pyridine-6-boronic acid

Precaución: For research use only.

Formula: C₇H₇BN₂O₂
MW: 161.9537 g/mol
MDL Number: MFCD06739225
CAS Number: 913835-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR