N-Propargyltrifluoroacetamide 98%, Ambeed.Inc

Supplier: Ambeed
Danger

Synonyms: 2,2,2-Trifluoro-N-(prop-2-yn-1-yl)acetamide, 2,2,2-Trifluoro-N-(2-propyn-1-yl)acetamide

A174341-5G A174341-1G A174341-250MG
77567-574EA 53 USD
77567-574 77611-696 77683-401
N-Propargyltrifluoroacetamide 98%, Ambeed.Inc
N-Propargyltrifluoroacetamide

Precaución: For research use only.

Formula: C₅H₄F₃NO
MW: 151.09 g/mol
MDL Number: MFCD14638370
CAS Number: 14719-21-2
UN: 1993
ADR: 3 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR