Diethylstilbestrol 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
(E)-3,4-Bis(4-hydroxyphenyl)-3-hexene, Stilbestrol, DES, (E)-4,4'-(Hex-3-ene-3,4-diyl)diphenol
A174248-25G
A174248-1G
A174248-100G
A174248-250MG
A174248-5G
77567-572EA
135.96
USD
77567-572
77600-416
77625-820
77625-822
77625-824
Diethylstilbestrol 98%
Diethylstilbestrol
Precaución: For research use only.
Formula:
C₁₈H₂₀O₂ MW: 268.356 g/mol |
MDL Number:
MFCD00002373 CAS Number: 56-53-1 UN: 3077 ADR: 9,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...