tert-Butyl (26-bromo-3,6,9,12,15,18,21,24-octaoxahexacosyl)carbamate 95%

Supplier: Ambeed
Warning

Synonyms: Boc-NH-PEG8-C2-Br

A1722039-1G A1722039-100MG A1722039-250MG
77567-532EA 577.83 USD
77567-532 77625-792 77625-794
tert-Butyl (26-bromo-3,6,9,12,15,18,21,24-octaoxahexacosyl)carbamate 95%
tert-Butyl (26-bromo-3,6,9,12,15,18,21,24-octaoxahexacosyl)carbamate

Precaución: For research use only.

Formula: C₂₃H₄₆BrNO₁₀
MW: 576.516 g/mol
CAS Number: 2688072-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR