4-(4-Chlorophenyl)pyrrolidin-2-one 97%

Supplier: Ambeed
Warning

A171443-5G A171443-1G
77567-524EA 758.08 USD
77567-524 77683-393
4-(4-Chlorophenyl)pyrrolidin-2-one 97%
4-(4-Chlorophenyl)pyrrolidin-2-one

Precaución: For research use only.

Formula: C₁₀H₁₀ClNO
MW: 195.65 g/mol
MDL Number: MFCD00854139
CAS Number: 22518-27-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR