(6-Cyano-5-fluoropyridin-3-yl)boronic acid 95%

Supplier: Ambeed
Warning

A1704646-1G A1704646-100MG A1704646-250MG
77567-498EA 241 USD
77567-498 77876-729 77876-730
(6-Cyano-5-fluoropyridin-3-yl)boronic acid 95%
(6-Cyano-5-fluoropyridin-3-yl)boronic acid

Precaución: For research use only.

Formula: C₆H₄BFN₂O₂
MDL Number: MFCD18260336
CAS Number: 2407353-18-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR