Dodecyl and tetradecyl glycidyl ethers

Supplier: Ambeed
Warning

A168935-1000G A168935-100G
77567-480EA 37.08 USD
77567-480 77567-482
Dodecyl and tetradecyl glycidyl ethers
Dodecyl and tetradecyl glycidyl ethers

Precaución: For research use only.

MDL Number: MFCD00192120
CAS Number: 68609-97-2

Order Now

Specification Test Results

Epoxy Value 0.32-0.37
Viscosity 4-10 cps

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR