2-Methyl-2-(6-oxo-1,6-dihydropyridin-2-yl)propanoic acid 97%
Supplier: Ambeed
|
Warning
|
A1681184-100MG
A1681184-50MG
77567-456EA
253
USD
77567-456
77593-026
2-Methyl-2-(6-oxo-1,6-dihydropyridin-2-yl)propanoic acid 97%
2-Methyl-2-(6-oxo-1,6-dihydropyridin-2-yl)propanoic acid
Precaución: For research use only.
Formula:
C₉H₁₁NO₃ MW: 181.189 g/mol |
CAS Number:
1785560-03-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...