tert-Butyl imidazolidine-1-carboxylate 95%

Supplier: Ambeed
Warning

A167789-100MG A167789-1G
77567-448EA 118.45 USD
77567-448 77567-450
tert-Butyl imidazolidine-1-carboxylate 95%
tert-Butyl imidazolidine-1-carboxylate

Precaución: For research use only.

Formula: C₈H₁₆N₂O
MW: 172.23 g/mol
MDL Number: MFCD22199572
CAS Number: 916891-97-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR