3-(diphenylphosphino)-propanoic acid 98%

Supplier: Ambeed
Warning

A167368-1G A167368-100MG A167368-250MG A167368-5G
77567-436EA 176.13 USD
77567-436 77615-392 77615-394 77615-396
3-(diphenylphosphino)-propanoic acid 98%
3-(diphenylphosphino)-propanoic acid

Precaución: For research use only.

Formula: C₁₅H₁₅O₂P
MW: 258.25 g/mol
MDL Number: MFCD00775376
CAS Number: 2848-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR