Poly-(L-lysine) 95%

Supplier: Ambeed
Warning

A165756-25G A165756-1G
77567-410EA 48.41 USD
77567-410 77667-320
Poly-(L-lysine) 95%
Poly-(L-lysine)

Precaución: For research use only.

Formula: (C₆H₁₄N₂O₂)x
MDL Number: MFCD01756704
CAS Number: 25104-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR