2,3,4,5,6-Penta(9H-carbazol-9-yl)benzonitrile 98%

Supplier: Ambeed
Warning

A1628080-100MG A1628080-250MG A1628080-1G
77567-376EA 200 USD
77567-376 77625-588 77876-447
2,3,4,5,6-Penta(9H-carbazol-9-yl)benzonitrile 98%
2,3,4,5,6-Penta(9H-carbazol-9-yl)benzonitrile

Precaución: For research use only.

Formula: C₆₇H₄₀N₆
CAS Number: 1469700-24-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR