5-Ethyl-2-hydroxybenzenecarbaldehyde 95%

Supplier: Ambeed
Warning

Synonyms: 5-Ethyl-2-hydroxybenzaldehyde

A162022-250MG A162022-1G
77567-366EA 111 USD
77567-366 77746-107
5-Ethyl-2-hydroxybenzenecarbaldehyde 95%
5-Ethyl-2-hydroxybenzenecarbaldehyde

Precaución: For research use only.

Formula: C₉H₁₀O₂
MDL Number: MFCD06739514
CAS Number: 52411-35-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR