6-Bromo-3,3-dimethylindolin-2-one 95%

Supplier: Ambeed
Warning

A160992-1G A160992-5G
77567-344EA 93.73 USD
77567-344 77685-319
6-Bromo-3,3-dimethylindolin-2-one 95%
6-Bromo-3,3-dimethylindolin-2-one

Precaución: For research use only.

Formula: C₁₀H₁₀BrNO
MW: 240.09 g/mol
MDL Number: MFCD18374762
CAS Number: 158326-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR