3-((4-Bromophenyl)amino)piperidine-2,6-dione 95%

Supplier: Ambeed
Warning

A1600540-250MG A1600540-100MG
77567-332EA 245 USD
77567-332 77876-351
3-((4-Bromophenyl)amino)piperidine-2,6-dione 95%
3-((4-Bromophenyl)amino)piperidine-2,6-dione

Precaución: For research use only.

Formula: C₁₁H₁₁BrN₂O₂
CAS Number: 2828444-36-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR