Aminoacetonitrile sulfate 98%

Supplier: Ambeed
Danger

A158694-5G A158694-25G
77567-306EA 6 USD
77567-306 77877-048
Aminoacetonitrile sulfate 98%
Aminoacetonitrile sulfate

Precaución: For research use only.

Formula: C₄H₁₀N₄O₄S
MDL Number: MFCD00012851
CAS Number: 5466-22-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR