1,1',1''-(1,3,5-Triazinane-1,3,5-triyl)tris(2-bromoethan-1-one) 95%

Supplier: Ambeed
Danger

A1576802-250MG A1576802-1G
77567-292EA 286 USD
77567-292 77876-321
1,1',1''-(1,3,5-Triazinane-1,3,5-triyl)tris(2-bromoethan-1-one) 95%
1,1',1''-(1,3,5-Triazinane-1,3,5-triyl)tris(2-bromoethan-1-one)

Precaución: For research use only.

Formula: C₉H₁₂Br₃N₃O₃
CAS Number: 92531-02-7
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR